Please login to your online account to display your discounted pricing

n-Butyllithium, 2.2M in hexane, packaged under Argon in resealable ChemSeal™ bottles, Alfa Aesar™

$34.64 - $34.64

Chemical Identifiers

Chemical Name or Material n-Butyllithium
CAS 109-72-8
Molecular Formula C4H9Li
Linear Formula CH3(CH2)3Li
Formula Weight 64.06
MDL Number MFCD00009414
UN Number UN3394
Beilstein 1209227
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA412483Y

 
alfa aesar
412483Y
Packaged under Argon in resealable ChemSeal™ bottles 0.5 mol CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41248JP

 
alfa aesar
41248JP
Packaged under Argon in resealable ChemSeal™ bottles 0.05 mol Each for $34.64
Description & Specifications

Specifications

Formula Weight 64.06
Quantity 0.5 mol
Chemical Name or Material n-Butyllithium
Concentration 2.2M in hexane
TSCA TSCA
Physical Form Liquid
Molecular Formula C4H9Li
Linear Formula CH3(CH2)3Li
MDL Number MFCD00009414
CAS 109-72-8
UN Number UN3394
Beilstein 1209227
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H250-H260-H252-H304-H361-H373-H314-H336;
Highly flammable liquid and vapour.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities; may catch fire.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Colorless to Pale Yellow
Flash Point −21°C (−7°F)
Odor Petroleum-like
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
Packaging Packaged under Argon in resealable ChemSeal™ bottles
Sensitivity Air and Moisture Sensitive
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H250-H260-H252-H304-H361-H373-H314-H336;
Highly flammable liquid and vapour.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities; may catch fire.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.