Please login to your online account to display your discounted pricing

n-Butyllithium, 2.2M in hexane, under Argon in resealable ChemSeal bottles, Alfa Aesar

Chemical Identifiers

Chemical Name or Material n-Butyllithium
CAS 109-72-8
Molecular Formula CH3(CH2)3Li
Formula Weight 64.06
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41248JR

  SDS
alfa aesar
41248JR
0.2 mole CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41248JT

  SDS
alfa aesar
41248JT
0.5 mole CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 64.06
Quantity 0.2 mole
Chemical Name or Material n-Butyllithium
Molecular Formula CH3(CH2)3Li
CAS 109-72-8