Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Butyllithium, 1.6M in hexanes, packaged under Argon in resealable ChemSeal™ bottles

Chemical Identifiers

Chemical Name or Material n-Butyllithium
CAS 109-72-8
Linear Formula CH3(CH2)3Li
Formula Weight 64.06
MDL Number MFCD00009414
UN Number UN3394
Beilstein 1209227
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA47107AE

  SDS
alfa aesar™
47107AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA47107AP

  SDS
alfa aesar™
47107AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 64.06
TSCA TSCA
Color Yellow to orange
Packaging ChemSeal™ bottles
Chemical Name or Material n-Butyllithium
Concentration 1.6M in hexanes
Physical Form Liquid
Linear Formula CH3(CH2)3Li
MDL Number MFCD00009414
CAS 109-72-8
Quantity 100mL
UN Number UN3394
Beilstein 1209227
Density 0.680g/mL
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H250-H260-H252-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities; may catch fire
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Packaged under Argon
DOT Information Transport Hazard Class: 4.2; Packaging Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
Sensitivity Air & Moisture Sensitive

  • Packaged under Argon

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H250-H260-H252-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities; may catch fire
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.

  • Packaged under Argon