missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Butyllithium, 1.6M in hexanes, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 47107AP

 View more versions of this product

Catalog No. AA47107AP



Description

Description

  • Packaged under Argon
Specifications

Specifications

n-Butyllithium
109-72-8
0.680g/mL
ChemSeal™ bottles
C4H9Li
MFCD00009414
1209227
DLEDOFVPSDKWEF-UHFFFAOYSA-N
lithium;butane
61028
Liquid
1.6M in hexanes
500mL
Yellow to Orange
Air & Moisture Sensitive
CH3(CH2)3Li
UN3394
n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc
[Li+].CCC[CH2-]
64.056
64.06

Chemical Identifiers

109-72-8
64.056
DLEDOFVPSDKWEF-UHFFFAOYSA-N
61028
[Li+].CCC[CH2-]
C4H9Li
MFCD00009414
n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc
lithium;butane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H250-H260-H252-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities; may catch fire
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packaging Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

SDS Documents