missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Butylimidazole, 98%, Acros Organics™

Manufacturer:  Acros Organics AC442771000

 View more versions of this product

Catalog No. AC442771000



Specifications

Specifications

N-Butylimidazole
113°C
0.5% max.
100g
C7H12N2
Solubility in water: soluble.
CCCCN1C=CN=C1
124.187
124.19
0.9450g/mL
Authentic
257.0°C
4316-42-1
1-butyl-1h-imidazole, 1h-imidazole, 1-butyl, n-butylimidazole, 1-n-butylimidazole, n-n-butyl imidazole, n-n-butyl imidazole, 1-n-butyl imidazole, 1-buthylimidazole, n-n-butylimidazole, imidazole, 1-butyl
MCMFEZDRQOJKMN-UHFFFAOYSA-N
1-butylimidazole
61347
98%

Chemical Identifiers

4316-42-1
124.187
1-butyl-1h-imidazole, 1h-imidazole, 1-butyl, n-butylimidazole, 1-n-butylimidazole, n-n-butyl imidazole, n-n-butyl imidazole, 1-n-butyl imidazole, 1-buthylimidazole, n-n-butylimidazole, imidazole, 1-butyl
1-butylimidazole
C7H12N2
MCMFEZDRQOJKMN-UHFFFAOYSA-N
61347
CCCCN1C=CN=C1
Safety and Handling

Safety and Handling

GHS H Statement Fatal if inhaled. Causes skin irritation. Causes serious eye damage. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Do not breathe dust/fume/gas/mist/vapors/spray.

Danger

SDS Documents