missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
n-Butylidenephthalide, (E)+(Z), 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1035330
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
n-Butylidenephthalide | |
551-08-6 | |
108°C to 109°C (0.1 mmHg) | |
C12H12O2 | |
MFCD00047319,MFCD00047319 | |
5145 | |
WMBOCUXXNSOQHM-DHZHZOJOSA-N | |
(3E)-3-butylidene-2-benzofuran-1-one | |
5352899 | |
95% |
(E)+(Z) | |
1.103 | |
132°C (269°F) | |
1.577 | |
250 g | |
ligusticum lactone, butylidene phthalide, n-butylidene phthalide, 3-butylidene-1 3h-isobenzofuranone, 3-butylidenephthalide, phthalide, 3-butylidene, fema no. 3333, bdph, 3-butylidene phthalide, 1 3h-isobenzofuranone, 3-butylidene | |
CCC\C=C1\OC(=O)C2=CC=CC=C12 | |
188.23 | |
188.23 |
Chemical Identifiers
551-08-6 | |
188.23 | |
WMBOCUXXNSOQHM-DHZHZOJOSA-N | |
5352899 | |
CCC\C=C1\OC(=O)C2=CC=CC=C12 |
C12H12O2 | |
MFCD00047319,MFCD00047319 | |
ligusticum lactone, butylidene phthalide, n-butylidene phthalide, 3-butylidene-1 3h-isobenzofuranone, 3-butylidenephthalide, phthalide, 3-butylidene, fema no. 3333, bdph, 3-butylidene phthalide, 1 3h-isobenzofuranone, 3-butylidene | |
(3E)-3-butylidene-2-benzofuran-1-one |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 208-991-3
RTECSNumber : TI3692500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only