Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Butylferrocene, 98%

$31.69 - $31.69

Chemical Identifiers

Chemical Name or Material n-Butylferrocene
CAS 31904-29-7
Molecular Formula C14H18Fe
Formula Weight 242.15g/mol
MDL Number MFCD00001435
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1671909

 
alfa aesar™
A16719-09
10g Each for $31.69
Description & Specifications

Specifications

Boiling Point 232°C (630mmHg)
Quantity 10g
Flash Point >110°C (230°F)
Chemical Name or Material n-Butylferrocene
CAS 31904-29-7
Assay 98%
Molecular Formula C14H18Fe
Formula Weight 242.15g/mol
MDL Number MFCD00001435
TSCA TSCA