Please login to your online account to display your discounted pricing

n-Butylethylmagnesium, 0.9M Solution in Heptane, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC381181000

 View more versions of this product

Catalog No. AC381181000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 110.48
Quantity 100mL
Color Clear colorless to slightly hazy
Packaging AcroSeal™ Glass Bottle
Density 0.7300g/mL
Linear Formula CH3CH2Mg(CH2)3CH3
Name Note 0.9M Solution in Heptane
Molecular Formula C6H14Mg
CAS 62202-86-2
CAS 142-82-5
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact
Chemical Name or Material n-Butylethylmagnesium
MDL Number MFCD00072569
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement May cause drowsiness or dizziness. Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. Very toxic to aquatic life with long lasting effects. In contact with water rel

Specifications

Chemical Name or Material n-Butylethylmagnesium
Name Note 0.9M Solution in Heptane
CAS 62202-86-2
CAS 142-82-5
Molecular Formula C6H14Mg
Linear Formula CH3CH2Mg(CH2)3CH3
Formula Weight 110.48
MDL Number MFCD00072569
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact

GHS P Statement May cause drowsiness or dizziness. Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. Very toxic to aquatic life with long lasting effects. In contact with water rel

SDS & Safety Documents