Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Butyldeoxynojirimycin, 98%


An →→alpha-glucosidase Inhibitor

Quantity 5mg

Manufacturer: alfa aesar™  J66613LB0

 View more versions of this product

Catalog No. AAJ66613LB0

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 219.28
Melting Point 126° to 127°C
Chemical Name or Material N-Butyldeoxynojirimycin
Physical Form Powder
Assay 98%
Molecular Formula C10H21NO4
MDL Number MFCD00272581
CAS 72599-27-0
Solubility Information Soluble in water at 10mg/mL
Color White
Storage Note 1 2° to 8°C
Quantity 5mg
Merck Index 14,6188
Recommended Storage 2° to 8°C
RTECS Number TN4350150

Specifications

Chemical Name or Material N-Butyldeoxynojirimycin
CAS 72599-27-0
Molecular Formula C10H21NO4
Formula Weight 219.28
MDL Number MFCD00272581
Merck Index 14,6188
Accessories

2° to 8°C

2° to 8°C