missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Butylbenzylamine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC306710100

 View more versions of this product

Catalog No. AC306710100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

N-Butylbenzylamine
2403-22-7
100.0
Authentic
1.5000 to 1.5020 (20°C, 589nm)
Liquid
10g
C11 H17 N
163.27
98%
97.5
0.9110g/mL
Glass bottle
Clear colorless to slightly yellow
≥97.5%
98%
C6H5CH2NH(CH2)3CH3
MFCD00009427

Chemical Identifiers

2403-22-7
100.0
C11 H17 N
163.27
N-Butylbenzylamine, 98%
97.5
98%
C6H5CH2NH(CH2)3CH3
MFCD00009427
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 219-287-0

TSCA : TSCA