Please login to your online account to display your discounted pricing

N-Butylbenzylamine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Butylbenzylamine
Name Note 98%
CAS 2403-22-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11 H17 N
Linear Formula C6H5CH2NH(CH2)3CH3
Formula Weight 163.27
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC306710100

  SDS
thermo fisher scientific
AC306710100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC306710500

  SDS
thermo fisher scientific
306710500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 163.27
Quantity 10g
Color Clear colorless to slightly yellow
EINECS Number 219-287-0
TSCA TSCA
Density 0.9110g/mL
CAS Max % 100.0
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C11 H17 N
Linear Formula C6H5CH2NH(CH2)3CH3
CAS 2403-22-7
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Chemical Name or Material N-Butylbenzylamine
Refractive Index 1.5000 to 1.5020 (20°C, 589nm)
Name Note 98%
MDL Number MFCD00009427
CAS Min % 97.5
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.