Learn More
n-Butyl vinyl ether, 98%, stab. with 0.01% KOH, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B23673AE
Description
Butyl Vinyl Ether is a reagent used in organic chemical reactions and the preparation of pharmaceutical compounds used in anticancer therapy. It is also used in the synthesis of potent inhibitors for PDE5. It is also used in the synthesis of copolymers and as an acrylic/vinyl acetate resin comonomer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water 3 g/L (20°C). Soluble in chloroform and methanol.
Notes
Store away from air and oxidizing agents may form peroxides. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Donot distill to dryness. Keep it refrigerated. Check the container pressure periodically to prevent peroxide formation.
Specifications
n-Butyl vinyl ether | |
111-34-2 | |
0.778 | |
−12°C (10°F) | |
1.401 | |
MFCD00009454 | |
UN3271 | |
n-butyl vinyl ether, butyl vinyl ether, vinyl butyl ether, butane, 1-ethenyloxy, butoxyethylene, 1-ethenyloxy butane, butoxyethene, ether, butyl vinyl, vinyl n-butyl ether, ethenyl n-butyl ether | |
UZKWTJUDCOPSNM-UHFFFAOYSA-N | |
1-ethenoxybutane | |
8108 | |
98% |
stabilized with 0.01% KOH | |
-113°C | |
92°C to 94°C | |
C6H12O | |
CH3(CH2)3OCH=CH2 | |
100 mL | |
1560217 | |
Soluble in water 3g/L (20°C). Soluble in chloroform and methanol. | |
CCCCOC=C | |
100.16 | |
100.16 |
Chemical Identifiers
111-34-2 | |
100.16 | |
UZKWTJUDCOPSNM-UHFFFAOYSA-N | |
8108 | |
CCCCOC=C |
C6H12O | |
MFCD00009454 | |
n-butyl vinyl ether, butyl vinyl ether, vinyl butyl ether, butane, 1-ethenyloxy, butoxyethylene, 1-ethenyloxy butane, butoxyethene, ether, butyl vinyl, vinyl n-butyl ether, ethenyl n-butyl ether | |
1-ethenoxybutane |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber : 203-860-7
RTECSNumber : KV5950000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only