Please login to your online account to display your discounted pricing

n-Butyl vinyl ether, 98%, ACROS Organics™

$47.94 - $47.94

Chemical Identifiers

Chemical Name or Material n-Butyl vinyl ether
Name Note Stabilized
CAS 111-34-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H12O
Linear Formula CH3(CH2)3OCH=CH2
Formula Weight 100.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC149705000

  SDS
thermo fisher scientific
149705000
Glass bottle 500mL Each for $47.94
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 100.16
Quantity 500mL
Color Clear
UN Number 2352
CAS Max % 100.0
Chemical Name or Material n-Butyl vinyl ether
Infrared Spectrum Authentic
Name Note Stabilized
Assay Percent Range 98%
Molecular Formula C6H12O
Linear Formula CH3(CH2)3OCH=CH2
Packaging Glass bottle
MDL Number MFCD00009454
CAS 111-34-2
Refractive Index 1.4005 to 1.4025 (20°C, 589nm)
Stabilizer max. 1000ppm KOH
Synonym 1-(Ethenyloxy)butane
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Extremely flammable liquid and vapour.
Causes skin irritation.
Butanol 0.5% max. (GC)
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge