Please login to your online account to display your discounted pricing

n-Butyl vinyl ether, 98%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  149705000

 View more versions of this product

Catalog No. AC149705000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 100.16
Quantity 500mL
Color Clear
UN Number 2352
CAS Max % 100.0
Chemical Name or Material n-Butyl vinyl ether
Infrared Spectrum Authentic
Name Note Stabilized
Assay Percent Range 98%
Molecular Formula C6H12O
Linear Formula CH3(CH2)3OCH=CH2
Packaging Glass bottle
MDL Number MFCD00009454
CAS 111-34-2
Refractive Index 1.4005 to 1.4025 (20°C, 589nm)
Stabilizer max. 1000ppm KOH
Synonym 1-(Ethenyloxy)butane
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Extremely flammable liquid and vapour.
Causes skin irritation.
Butanol 0.5% max. (GC)
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

Specifications

Chemical Name or Material n-Butyl vinyl ether
Name Note Stabilized
CAS 111-34-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H12O
Linear Formula CH3(CH2)3OCH=CH2
Formula Weight 100.16
MDL Number MFCD00009454
Synonym 1-(Ethenyloxy)butane
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

GHS H Statement:
Extremely flammable liquid and vapour.
Causes skin irritation.

SDS & Safety Documents