missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(n-Butyl)triphenylphosphonium Bromide 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 108061000

 View more versions of this product

Catalog No. AC108061000


 

Specifications Description & Specifications

Specifications

n-Butyltriphenylphosphonium bromide
1779-51-7
100.0
Crystalline Powder
99%
CH3(CH2)3P(C6H5)3Br
399.3
240° to 243°C
100g
99%
98.5
Authentic
≥98.5%
C22H24BrP
Glass bottle
MFCD00011855
White

Chemical Identifiers

1779-51-7
100.0
C22H24BrP
399.3
n-Butyltriphenylphosphonium bromide, 99%
98.5
99%
CH3(CH2)3P(C6H5)3Br
MFCD00011855
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-219-4

RTECSNumber : TA1855200

TSCA : TSCA