Please login to your online account to display your discounted pricing

n-Butyl sulfide, 99%, pure, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: fisher scientific acros  108011000

 View more versions of this product

Catalog No. AC108011000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.5%
Formula Weight 146.29
Quantity 100mL
Color Clear colorless to very slightly yellow
Purity Grade Pure
Density 0.8400g/mL
Chemical Name or Material n-Butyl sulfide
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula [CH3(CH2)3]2S
Molecular Formula C8H18S
Packaging Glass bottle
CAS 544-40-1
Refractive Index 1.4520 to 1.4540 (20°C, 589nm)
Synonym Di-n-butyl sulfide

Specifications

Chemical Name or Material n-Butyl sulfide
CAS 544-40-1
Purity Grade Pure
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H18S
Linear Formula [CH3(CH2)3]2S
Formula Weight 146.29
Synonym Di-n-butyl sulfide