missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Butyl phosphate, mixture of mono-n-butyl and di-n-butyl, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 022404A1

 View more versions of this product

Catalog No. AA22404A1



Description

Description

n-Butyl phosphate is a solvent and plasticizer for cellulose esters such as nitrocellulose and cellulose acetate. It forms stable hydrophobic complexes with some metals, as an extractant. It is also used in industry are as a component of aircraft hydraulic fluid and as a solvent for extraction and purification of rare earth metals from their ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Butyl phosphate
52933-01-4
Colorless
>124°C (decomposition)
C4H11O4P
MFCD00013947
UN1718
Soluble in ethanol,diethyl ether. Insoluble in water.
CCCCOP(=O)(O)O
154.102
Liquid
Mixture of mono-n-butyl and di-n-butyl
-60°C
1.123 g/mL
110°C (230°F)
1.429
1 kg
n-butylphosphate, monobutyl phosphate, phosphoric acid, monobutyl ester, butyl diacid phosphate, n-butyl acid phosphate, butyl dibasic acid phosphate, unii-0i9w48e53l, butyl phosphate, buo ho 2po, mono-n-butylphosphoric acid, butoxyphosphonic acid
BNMJSBUIDQYHIN-UHFFFAOYSA-N
butyl dihydrogen phosphate
15389

Chemical Identifiers

52933-01-4
154.102
BNMJSBUIDQYHIN-UHFFFAOYSA-N
15389
CCCCOP(=O)(O)O
C4H11O4P
MFCD00013947
n-butylphosphate, monobutyl phosphate, phosphoric acid, monobutyl ester, butyl diacid phosphate, n-butyl acid phosphate, butyl dibasic acid phosphate, unii-0i9w48e53l, butyl phosphate, buo ho 2po, mono-n-butylphosphoric acid, butoxyphosphonic acid
butyl dihydrogen phosphate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335-H351

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: BUTYL ACID PHOSPHATE

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only