missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Butyl Phosphate (Laboratory), Fisher Chemical™

$218.69 - $1,575.15

Chemical Identifiers

CAS 126-73-8
Molecular Formula C12H27O4P
Molecular Weight (g/mol) 266.318
MDL Number MFCD00009436
InChI Key STCOOQWBFONSKY-UHFFFAOYSA-N
Synonym butyl phosphate, celluphos 4, disflamoll tb, phosphoric acid tributyl ester, tri-n-butyl phosphate, tributilfosfato, tributyle phosphate de, tributylfosfaat, tributylphosphat, tributylphosphate
PubChem CID 31357
ChEBI CHEBI:35019
IUPAC Name tributyl phosphate
SMILES CCCCOP(=O)(OCCCC)OCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
B404-500
SDS Accessories View Documents
Fisher Chemical
BPB404500
500mL Glass Bottle Each for $218.69
Case of 6 Each for $1,312.14
 
B404-4
SDS View Documents
Fisher Chemical
BPB4044
4L Glass Bottle Each for $1,575.15
Case of 4 Each for $6,300.60
 
Description

Description

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Specifications

Chemical Identifiers

126-73-8
266.318
STCOOQWBFONSKY-UHFFFAOYSA-N
31357
tributyl phosphate
C12H27O4P
MFCD00009436
butyl phosphate, celluphos 4, disflamoll tb, phosphoric acid tributyl ester, tri-n-butyl phosphate, tributilfosfato, tributyle phosphate de, tributylfosfaat, tributylphosphat, tributylphosphate
CHEBI:35019
CCCCOP(=O)(OCCCC)OCCCC

Specifications

0.979g/cm³
Glass Bottle
Colorless
500mL
C12H27O4P
MFCD00009436
STCOOQWBFONSKY-UHFFFAOYSA-N
tributyl phosphate
31357
266.32
Laboratory
Pass Test
289°C
-79°C
126-73-8
(C4H9)3PO4
butyl phosphate, celluphos 4, disflamoll tb, phosphoric acid tributyl ester, tri-n-butyl phosphate, tributilfosfato, tributyle phosphate de, tributylfosfaat, tributylphosphat, tributylphosphate
CCCCOP(=O)(OCCCC)OCCCC
266.318
CHEBI:35019
Liquid
n-Butyl Phosphate
Safety and Handling

Safety and Handling