Please login to your online account to display your discounted pricing

n-Butyl Phenyl Ether, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material n-Butyl phenyl ether
CAS 1126-79-0
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C10H14O
Linear Formula C6H5O(CH2)3CH3
Formula Weight 150.22g/mol
MDL Number MFCD00009438
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC161921000

  SDS
fisher scientific acros
161921000
Glass bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4960 to 1.4980 (20°C, 589nm)
Color Clear colorless
Quantity 100mL
Chemical Name or Material n-Butyl phenyl ether
CAS 1126-79-0
CAS Min % 98.5
CAS Max % 100.0
Physical Form Clear colorless liquid
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C10H14O
Linear Formula C6H5O(CH2)3CH3
Formula Weight 150.22g/mol
MDL Number MFCD00009438
Beilstein 06, 143
Synonym n-Butoxybenzene