Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Butyl-N-ethylethane-1,2-diamine

$33.59 - $146.16

Chemical Identifiers

Chemical Name or Material N-Butyl-N-ethylethane-1,2-diamine
CAS 19435-65-5
Molecular Formula C8H20N2
Formula Weight 144.26
MDL Number MFCD00047953
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5032403

  SDS
alfa aesar™
H5032403
1g Each for $33.59

AAH5032406

  SDS
alfa aesar™
H5032406
5g Each for $146.16
Description & Specifications

Specifications

Formula Weight 144.26
CAS 19435-65-5
MDL Number MFCD00047953
Molecular Formula C8H20N2
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material N-Butyl-N-ethylethane-1,2-diamine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.