Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Butyl methyl sulfide, 98+%

Chemical Identifiers

Chemical Name or Material n-Butyl methyl sulfide
CAS 628-29-5
Molecular Formula C5H12S
Linear Formula CH3S(CH2)3CH3
Formula Weight 104.22
MDL Number MFCD00015256
UN Number UN1993
Beilstein 1730959
Synonym Methyl n-butyl sulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1203906

  SDS
alfa aesar™
L1203906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1203914

  SDS
alfa aesar™
L1203914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 104.22
Synonym Methyl n-butyl sulfide
Chemical Name or Material n-Butyl methyl sulfide
CAS 628-29-5
Health Hazard 3 GHS P Statement
P210-P261-P243
Keep away from heat/sparks/open flames/hot surfaces. - No Smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Take precautionary measures against static discharge.
MDL Number MFCD00015256
Assay ≥98%
TSCA TSCA
Boiling Point 122° to 123°C
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapor.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Health Hazard 1 Danger
Molecular Formula C5H12S
Flash Point 20°C (68°F)
Linear Formula CH3S(CH2)3CH3
Odor Unpleasant
Quantity 5g
Beilstein 1730959
UN Number UN1993
Safety and Handling

Danger

GHS P Statement
P210-P261-P243
Keep away from heat/sparks/open flames/hot surfaces. - No Smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Take precautionary measures against static discharge.

GHS H Statement
H225
Highly flammable liquid and vapor.