Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Butyl L-lactate, 97+%

$49.86 - $549.97

Chemical Identifiers

Chemical Name or Material n-Butyl L-lactate
CAS 34451-19-9
Molecular Formula C7H14O3
Formula Weight 146.19
MDL Number MFCD00004519
Synonym n-Butyl (S)-(-)-2-hydroxypropionate; L-Lactic acid n-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA138210B

  SDS
alfa aesar™
A138210B
1000g Each for $132.45

AAA138210I

  SDS
alfa aesar™
A138210I
5000g Each for $549.97

AAA1382130

  SDS
alfa aesar™
A1382130
250g Each for $49.86
Description & Specifications

Specifications

Formula Weight 146.19
Flash Point 69°C (157°F)
Molecular Formula C7H14O3
Boiling Point 185° to 187°C
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym n-Butyl (S)-(-)-2-hydroxypropionate; L-Lactic acid n-butyl ester
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Chemical Name or Material n-Butyl L-lactate
CAS 34451-19-9
Quantity 1000g
Assay ≥97%
TSCA TSCA
Health Hazard 1 Warning
MDL Number MFCD00004519

Applications
Fenofibrate was formulated with n-butyl L-lactate, Tween 80, and a number of cosurfactants, diluted with the water phase (either water or saccharidesolution) and then subjected to a freeze-drying (FD) process to obtain SMEPMS nanosized.

Solubility
Soluble in alcohol and water(42 g/L (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.