Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Butyl heptafluorobutyrate, 97%

$135.31 - $135.31

Chemical Identifiers

Chemical Name or Material n-Butyl heptafluorobutyrate
CAS 1559-07-5
Molecular Formula C8F7H9O2
Linear Formula CF3CF2CF2CO2(CH2)3CH3
Formula Weight 270.15
MDL Number MFCD00039240
Beilstein 1798327
Synonym Heptafluorobutyric acid n-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2240018

 
alfa aesar™
B2240018
50g Each for $135.31
Description & Specifications

Specifications

Formula Weight 270.15
Chemical Name or Material n-Butyl heptafluorobutyrate
Assay 97%
Molecular Formula C8F7H9O2
Linear Formula CF3CF2CF2CO2(CH2)3CH3
MDL Number MFCD00039240
CAS 1559-07-5
Boiling Point 131° to 133°C
Beilstein 1798327
Quantity 50g
Synonym Heptafluorobutyric acid n-butyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.