Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Butyl glycidyl ether, 95%

Chemical Identifiers

Chemical Name or Material n-Butyl glycidyl ether
CAS 2426-08-6
Molecular Formula C7H14O2
Formula Weight 130.19
MDL Number MFCD00005145
UN Number UN3271
Beilstein 103668
Synonym Butyl 2,3-epoxypropyl ether; Glycidol butyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL16201AD

  SDS
alfa aesar™
L16201AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL16201AK

  SDS
alfa aesar™
L16201AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 130.19
Chemical Name or Material n-Butyl glycidyl ether
CAS 2426-08-6
MDL Number MFCD00005145
Assay 95%
Health Hazard 2 GHS H Statement
H226-H341-H351-H302-H332-H317-H335
Flammable liquid and vapor.
Suspected of causing genetic defects.
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause respiratory irritation.
Quantity 50mL
Synonym Butyl 2,3-epoxypropyl ether; Glycidol butyl ether
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
Health Hazard 3 GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Pungent
Health Hazard 1 Warning
RTECS Number TX4200000
Flash Point 40°C (104°F)
Molecular Formula C7H14O2
UN Number UN3271
Boiling Point 163° to 165°C
Beilstein 103668
TSCA TSCA

Applications
n-Butyl glycidyl ether is used as a casting material, adhesive and as an epoxy thinner.

Solubility
Soluble in water (20g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to moisture. Incompatible with oxidizing agents, acids, bases, alcohols and amines.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H341-H351-H302-H332-H317-H335
Flammable liquid and vapor.
Suspected of causing genetic defects.
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause respiratory irritation.