Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Butyl formate, 97%

Chemical Identifiers

Chemical Name or Material n-Butyl formate
CAS 592-84-7
Molecular Formula HCO2(CH2)3CH3
Formula Weight 102.13
MDL Number MFCD00003296
UN Number UN1128
Beilstein 1742108
Synonym Formic acid n-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA13882AK

  SDS
alfa aesar™
A13882AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 102.13
Chemical Name or Material n-Butyl formate
TSCA TSCA
Assay 97%
Molecular Formula HCO2(CH2)3CH3
Melting Point −92°C
MDL Number MFCD00003296
CAS 592-84-7
UN Number UN1128
Boiling Point 105° to 107°C
Beilstein 1742108
Quantity 250mL
Flash Point 18°C (64°F)
Synonym Formic acid n-butyl ester
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H319-H335
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: N-BUTYL FORMATE
RTECS Number LQ5500000
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H319-H335
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.