Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Butyl ethyl ether, 98+%

Chemical Identifiers

Chemical Name or Material n-Butyl ethyl ether
CAS 628-81-9
Molecular Formula CH3CH2CH2CH2OCH2CH3
Formula Weight 102.18
MDL Number MFCD00009455
UN Number UN1179
Beilstein 1731323
Synonym 1-Ethoxybutane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1796309

  SDS
alfa aesar™
A1796309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 102.18
Flash Point −6°C (21°F)
UN Number UN1179
Boiling Point 91° to 92°C
Health Hazard 2 GHS H Statement
H225-H301-H332
Highly flammable liquid and vapour.
Toxic if swallowed.
Harmful if inhaled.
Melting Point −124°C
RTECS Number KN4725000
MDL Number MFCD00009455
TSCA TSCA
Quantity 10g
Beilstein 1731323
Synonym 1-Ethoxybutane
Chemical Name or Material n-Butyl ethyl ether
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL BUTYL ETHER
Health Hazard 3 GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Solubility Information Slightly soluble in water
Assay ≥98%
Health Hazard 1 Danger
Molecular Formula CH3CH2CH2CH2OCH2CH3
CAS 628-81-9
Safety and Handling

Danger

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H301-H332
Highly flammable liquid and vapour.
Toxic if swallowed.
Harmful if inhaled.