Please login to your online account to display your discounted pricing

n-Butyl cyanoacetate, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material n-Butyl cyanoacetate
CAS 5459-58-5
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C7H11NO2
Linear Formula NCCH2C(O)OCH2CH2CH2CH3
Formula Weight 141.16
MDL Number MFCD00001941
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC248011000

  SDS
thermo fisher scientific
248011000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Liquid
Formula Weight 141.16
Quantity 100g
Color Clear colorless to light yellow
CAS Max % 100.0
CAS Min % 99.0
Packaging Glass bottle
Chemical Name or Material n-Butyl cyanoacetate
CAS 5459-58-5
Molecular Formula C7H11NO2
Assay Percent Range 99+%
MDL Number MFCD00001941
Refractive Index 1.4250 to 1.4270 (20°C, 589nm)
Infrared Spectrum Authentic
Linear Formula NCCH2C(O)OCH2CH2CH2CH3