Please login to your online account to display your discounted pricing

n-Butyl cyanoacetate, 99+%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  248011000

 View more versions of this product

Catalog No. AC248011000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Liquid
Formula Weight 141.16
Quantity 100g
Color Clear colorless to light yellow
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material n-Butyl cyanoacetate
Packaging Glass bottle
Assay Percent Range 99+%
Molecular Formula C7H11NO2
MDL Number MFCD00001941
CAS 5459-58-5
Refractive Index 1.4250 to 1.4270 (20°C, 589nm)
Linear Formula NCCH2C(O)OCH2CH2CH2CH3
Infrared Spectrum Authentic

Specifications

Chemical Name or Material n-Butyl cyanoacetate
CAS 5459-58-5
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C7H11NO2
Linear Formula NCCH2C(O)OCH2CH2CH2CH3
Formula Weight 141.16
MDL Number MFCD00001941