Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Butyl cinnamate, 98%

$236.57 - $236.57

Chemical Identifiers

Chemical Name or Material n-Butyl cinnamate
CAS 538-65-8
Molecular Formula C13H16O2
Formula Weight 204.27
MDL Number MFCD00051560
Beilstein 2328067
Merck Index 14,2299
Synonym Cinnamic acid n-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0736230

  SDS
alfa aesar™
L0736230
250g Each for $236.57
Description & Specifications

Specifications

Formula Weight 204.27
CAS 538-65-8
MDL Number MFCD00051560
Health Hazard 1 Warning
Beilstein 2328067
Quantity 250g
RTECS Number GD8425000
Molecular Formula C13H16O2
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Merck Index 14,2299
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Synonym Cinnamic acid n-butyl ester
Flash Point 90°C (194°F)
Boiling Point 143° to 144°C (12mmHg)
Chemical Name or Material n-Butyl cinnamate
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.