Please login to your online account to display your discounted pricing

n-Butyl Chloride (Certified), Fisher Chemical


Quantity 1L
Packaging Glass Bottle

Manufacturer: fisher scientific company  BPB4161

 View more versions of this product

Catalog No. B416-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Formula Weight 92.57
Boiling Point 77°C
Melting Point −123°C
Quantity 1L
UN Number 1127
Flash Point −12°C
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Vapor Pressure 108mbar at 20°C
Color Colorless
Density 0.880g/cm³
ChemAlert Storage Symbol Red
Packaging Glass Bottle
CAS 109-69-3
Linear Formula CH3(CH2)3Cl
Chemical Name or Material n-Butyl Chloride
MDL Number MFCD00001009
Molecular Formula C4H9Cl
Water 0.05% max.
Cleanup Symbol Solvent
Synonym 1-Chlorobutane
Identification Pass Test
Additional Information Vapor Pressure: 108mbar at 20°C
Boiling Range 2°C max.
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. The toxicological properties have not been fully investigated. Wear personal protective equipment. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools. Use explosion-proof equipment. To avoid ignition of vapors by static electricity discharge, all metal parts of the equipment must b. Take precautionary measures against static discharges. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:1
Flammability:3
Instability:0

Specifications

Chemical Name or Material n-Butyl Chloride
CAS 109-69-3
Purity Grade Certified Grade
Molecular Formula C4H9Cl
Linear Formula CH3(CH2)3Cl
Formula Weight 92.57
MDL Number MFCD00001009
Synonym 1-Chlorobutane
Accessories
 

Emergency Overview
Flammable liquid and vapor. The toxicological properties have not been fully investigated. Wear personal protective equipment. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools. Use explosion-proof equipment. To avoid ignition of vapors by static electricity discharge, all metal parts of the equipment must b. Take precautionary measures against static discharges. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!

 

SDS & Safety Documents