Learn More
n-Butyl acetate, Semiconductor Grade, 99% min, Thermo Scientific Chemicals
$96.97 - $332.37
Chemical Identifiers
CAS | 123-86-4 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00009445 |
InChI Key | DKPFZGUDAPQIHT-UHFFFAOYSA-N |
Synonym | n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle |
PubChem CID | 31272 |
ChEBI | CHEBI:31328 |
IUPAC Name | butyl acetate |
SMILES | CCCCOC(C)=O |
Description
n-Butyl acetate is widely used as a solvent for cellulose nitrate, resins, polymers, oils and fats. It is used in paints, coatings, printing inks, aerosol sprays and cosmetics. It is used as a synthetic fruit flavoring agent in foods like candy, ice cream, cheeses and baked food items. In the pharmaceutical industry, it is used for extraction and as processing solvent. It is an ingredient of cleaners, essences and fragrances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123-86-4 | |
116.16 | |
DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
31272 | |
butyl acetate |
C6H12O2 | |
MFCD00009445 | |
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle | |
CHEBI:31328 | |
CCCCOC(C)=O |
Specifications
123-86-4 | |
Colorless | |
5 | |
25°C (77°F) | |
C6H12O2 | |
CH3CO2(CH2)3CH3 | |
1 L | |
1741921 | |
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle | |
DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
butyl acetate | |
31272 | |
116.16 | |
Semiconductor | |
n-Butyl acetate |
-78°C | |
0.881 g/mL | |
124°C to 126°C | |
Fruit-like | |
1.3945 | |
MFCD00009445 | |
UN1123 | |
14,1535 | |
Miscible with ethanol,ethyl ether,acetone,chloroform and most hydrocarbons. Slightly miscible with water. | |
CCCCOC(C)=O | |
116.16 | |
CHEBI:31328 | |
≤99% | |
Liquid |
Safety and Handling
GHS H Statement
H226-H336
Flammable liquid and vapor.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYL ACETATES
EINECSNumber : 204-658-1
RTECSNumber : AF350000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only