missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Butyl acetate, Semiconductor Grade, 99% min

Manufacturer:  Alfa Aesar™ 19395M1

 View more versions of this product

Catalog No. AA19395M1



Specifications

Specifications

n-Butyl acetate
25° (77°F)
1.3945
Colorless
4 x 1L
123-86-4
CH3CO2(CH2)3CH3
UN1123
14,1535
DKPFZGUDAPQIHT-UHFFFAOYSA-N
butyl acetate
31272
116.16
≤99%
0.881g/mL
Fruit-like
124°C to 126°C
-78°C
5
C6H12O2
MFCD00009445
1741921
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
CCCCOC(=O)C
116.16
CHEBI:31328
Liquid
Semiconductor

Chemical Identifiers

123-86-4
116.16
DKPFZGUDAPQIHT-UHFFFAOYSA-N
31272
butyl acetate
C6H12O2
MFCD00009445
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
CHEBI:31328
CCCCOC(=O)C
Safety and Handling

Safety and Handling

GHS H Statement
H226-H336
Flammable liquid and vapor.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYL ACETATES

RTECSNumber : AF350000

TSCA : TSCA

SDS Documents