Please login to your online account to display your discounted pricing

n-Butyl Acetate (Reagent), Fisher Chemical


Quantity 1L
Packaging Glass Bottle

Manufacturer: fisher scientific company  BPB3961

 View more versions of this product

Catalog No. B396-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Melting Point −77.9°C
Formula Weight 116.16
Boiling Point 125°C
Purity Grade Reagent Grade
Quantity 1L
UN Number 1123
Flash Point 22°C
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Molecular Formula C6H12O2
Density 0.880g/cm³
Color Colorless
pH 6.2
Vapor Pressure 13mbar at 20°C
Packaging Glass Bottle
Boiling Range 5°C max.
Health Hazard 2 WARNING!
Cleanup Symbol Solvent
Linear Formula CH3COOCH2(CH2)2CH3
Chemical Name or Material n-Butyl Acetate
MDL Number MFCD00009445
CAS 123-86-4
Identification Pass Test
Additional Information Vapor Pressure: 13mbar at 20°C
Synonym Acetic Acid Butyl Ester
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. May cause pulmonary edema. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:3
Instability:0
ChemAlert Storage Symbol Red

Specifications

Chemical Name or Material n-Butyl Acetate
CAS 123-86-4
Purity Grade Reagent Grade
Molecular Formula C6H12O2
Linear Formula CH3COOCH2(CH2)2CH3
Formula Weight 116.16
MDL Number MFCD00009445
Synonym Acetic Acid Butyl Ester
Accessories
 

Emergency Overview
Flammable liquid and vapor. May cause pulmonary edema. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:3
Instability:0

WARNING!

 

SDS & Safety Documents