Learn More
n-Butyl acetate, 99+%, for spectroscopy, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 167680010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopySpecifications
n-Butyl acetate | |
123-86-4 | |
-78°C | |
127°C | |
99% min. (GC) | |
C6H12O2 | |
CH3CO2(CH2)3CH3 | |
1 L | |
0.0005% max. | |
15, 1537 | |
Solubility in water: 7g/L (20°C) | |
CCCCOC(C)=O | |
116.16 | |
0.69 mPa.s (25°C) | |
116.16 | |
99+% | |
Liquid |
99+% | |
(1 cm cell vs water), 0.01 max. at 320nm, 0.02 max. at 300nm, 0.04 max. at 275nm, 0.15 max. at 260nm, 0.4 max. at 256nm, 1 max. at 254nm | |
0.8800g/mL | |
27°C | |
Glass bottle | |
1.3930 to 1.395 | |
MFCD00009445 | |
02, 130 | |
0.88 | |
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle | |
DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
butyl acetate | |
31272 | |
CHEBI:31328 | |
13mbar at 20°C | |
Spectroscopy |
Chemical Identifiers
123-86-4 | |
116.16 | |
DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
31272 | |
butyl acetate |
C6H12O2 | |
MFCD00009445 | |
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle | |
CHEBI:31328 | |
CCCCOC(C)=O |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Avoid breathing dust/fume/gas/mist/vapors/spray.
GHS Signal Word: Warning
EINECSNumber : 204-658-1
RTECSNumber : AF7350000
TSCA : TSCA
RUO – Research Use Only