Please login to your online account to display your discounted pricing

n-Butyl acetate, 99+%, extra pure, ACROS Organics™


Quantity 1L
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  107750010

 View more versions of this product

Catalog No. AC107750010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 116.16
Purity Grade Extra Pure
Physical Form Liquid
Quantity 1L
Color Clear
Beilstein 02, 130
Merck Index 15, 1537
Packaging Glass Bottle
CAS Min % 99.0
CAS Max % 100.0
Residue after Evaporation 0.005% max.
Linear Formula CH3CO2(CH2)3CH3
Refractive Index 1.3930 to 1.3950 (20°C, 589nm)
Chemical Name or Material n-Butyl acetate
CAS 123-86-4
Water 0.1% max. (K.F.)
Assay Percent Range 99+%
Infrared Spectrum Authentic
Molecular Formula C6H12O2
Health Hazard 1 Warning
MDL Number MFCD00009445
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

Specifications

Chemical Name or Material n-Butyl acetate
CAS 123-86-4
CAS Min % 99.0
CAS Max % 100.0
Purity Grade Extra Pure
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C6H12O2
Linear Formula CH3CO2(CH2)3CH3
Formula Weight 116.16
MDL Number MFCD00009445
Accessories

Warning

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

SDS & Safety Documents