Learn More
n-Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
$147.88 - $147.88
Chemical Identifiers
CAS | 94-26-8 |
---|---|
Molecular Formula | C11H14O3 |
Molecular Weight (g/mol) | 194.23 |
MDL Number | MFCD00016478 |
InChI Key | QFOHBWFCKVYLES-UHFFFAOYSA-N |
Synonym | butylparaben, butyl paraben, butyl p-hydroxybenzoate, nipabutyl, butoben, butyl chemosept, butyl parasept, butyl tegosept, butyl butex, tegosept butyl |
PubChem CID | 7184 |
IUPAC Name | butyl 4-hydroxybenzoate |
SMILES | CCCCOC(=O)C1=CC=C(C=C1)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1404330
|
Thermo Scientific Chemicals
A1404330 |
250 g |
Each for $147.88
|
|
|||||
Description
It is used as an preservative in cosmetics such as eye shadow, foundation, sunscreen, facial moisturizer and skin anti-aging treatment. It is also used in medication suspensions, and as a flavoring additive in food.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an preservative in cosmetics such as eye shadow, foundation, sunscreen, facial moisturizer and skin anti-aging treatment. It is also used in medication suspensions, and as a flavoring additive in food.
Solubility
Soluble in DMSO, ethyl acetate, methanol.
Notes
Store in refrigerator. Store away from oxidizing agents.
Chemical Identifiers
94-26-8 | |
194.23 | |
QFOHBWFCKVYLES-UHFFFAOYSA-N | |
7184 | |
CCCCOC(=O)C1=CC=C(C=C1)O |
C11H14O3 | |
MFCD00016478 | |
butylparaben, butyl paraben, butyl p-hydroxybenzoate, nipabutyl, butoben, butyl chemosept, butyl parasept, butyl tegosept, butyl butex, tegosept butyl | |
butyl 4-hydroxybenzoate |
Specifications
94-26-8 | |
C11H14O3 | |
250 g | |
14,1584 | |
Soluble in DMSO,ethyl acetate,methanol. | |
CCCCOC(=O)C1=CC=C(C=C1)O | |
194.23 | |
194.23 | |
n-Butyl 4-hydroxybenzoate |
67°C to 71°C | |
MFCD00016478 | |
1103741 | |
butylparaben, butyl paraben, butyl p-hydroxybenzoate, nipabutyl, butoben, butyl chemosept, butyl parasept, butyl tegosept, butyl butex, tegosept butyl | |
QFOHBWFCKVYLES-UHFFFAOYSA-N | |
butyl 4-hydroxybenzoate | |
7184 | |
≥99% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 202-318-7
RTECSNumber : DH1980000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only