Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Butoxyacetic acid, 95%

$53.73 - $213.96

Chemical Identifiers

Chemical Name or Material n-Butoxyacetic acid
CAS 2516-93-0
Molecular Formula CH3CH2CH2CH2OCH2CO2H
Formula Weight 132.16
MDL Number MFCD00067044
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5021006

 
alfa aesar™
H5021006
5g Each for $213.96

AAH5021003

 
alfa aesar™
H5021003
1g Each for $53.73
Description & Specifications

Specifications

Formula Weight 132.16
CAS 2516-93-0
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material n-Butoxyacetic acid
Assay 95%
Molecular Formula CH3CH2CH2CH2OCH2CO2H
MDL Number MFCD00067044
Boiling Point 234°C
Quantity 5g
Health Hazard 2 GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.