missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Butanol (Clear, Colorless Liq.), Fisher BioReagents

$72.23 - $72.23

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP505-500 SDS View Documents Fisher BioReagents
BPBP505500
500mL Glass Bottle Each for $72.23
Case of 6 Each for $433.38
 
Description

Description

n-Butanol is used as an analytical reagent and solvent.
Specifications

Chemical Identifiers

71-36-3
74.123
1-butanol
CHEBI:28885
CCCCO
C4H10O
LRHPLDYGYMQRHN-UHFFFAOYSA-N
263
butan-1-ol

Specifications

71-36-3
Vapor Pressure: 6.7mbar at 20°C
≥99.8 %
CH3CH2CH2CH2OH
15, 1542
LRHPLDYGYMQRHN-UHFFFAOYSA-N
butan-1-ol
263
74.12
≥99.8%
0.810g/cm³
Glass Bottle
0.1% max.
Butanol
Colorless
500mL
0.005% max.
Not Detected Detected/Not Detect.
C4H10O
1120
1-butanol
CCCCO
74.123
CHEBI:28885
Liquid
Pass Test
0.1% max.
6.7mbar at 20°C
2.95 mPa/s at 20°C
117.6°C
-89°C
n-Butanol
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Flammable liquid and vapor. Risk of serious damage to eyes. Irritating to eyes, respiratory system and skin. Vapors may cause drowsiness and dizziness. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:2
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

Recommended Storage : RT