Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Bsmoc-L-tryptophan, 97%

$181.97 - $181.97

Chemical Identifiers

Chemical Name or Material N-Bsmoc-L-tryptophan
CAS 197245-27-5
Molecular Formula C21H18N2O6S
Formula Weight 426.45g/mol
MDL Number MFCD01090981
Beilstein 8289582
Synonym N-(Benzo[b]thiophenesulfone-2-methoxycarbonyl)-L-tryptophan; Bsmoc-Trp-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1974306

 
alfa aesar™
L19743-06
5g Each for $181.97
Description & Specifications

Specifications

Quantity 5g
Melting Point 80° to 106°C
Quantity 5g
Optical Rotation −28° (c=0.5 in DMF)
Chemical Name or Material N-Bsmoc-L-tryptophan
CAS 197245-27-5
Assay 97%
Molecular Formula C21H18N2O6S
Formula Weight 426.45g/mol
MDL Number MFCD01090981
Beilstein 8289582
Synonym N-(Benzo[b]thiophenesulfone-2-methoxycarbonyl)-L-tryptophan; Bsmoc-Trp-OH
Recommended Storage 2° to 8°C