Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Bsmoc-L-phenylalanine, 97%

$33.86 - $290.92

Chemical Identifiers

Chemical Name or Material N-Bsmoc-L-phenylalanine
CAS 197245-19-5
Molecular Formula C19H17NO6S
Formula Weight 387.41
MDL Number MFCD03792475
Beilstein 8282210
Synonym N-(Benzo[b]thiophenesulfone-2-methoxycarbonyl)-L-phenylalanine; Bsmoc-Phe-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1974106

 
alfa aesar™
L1974106
5g Each for $290.92

AAL1974103

 
alfa aesar™
L1974103
1g Each for $84.81

AAL19741MD

 
alfa aesar™
L19741MD
250mg Each for $33.86
Description & Specifications

Specifications

Formula Weight 387.41
Chemical Name or Material N-Bsmoc-L-phenylalanine
Assay 97%
Molecular Formula C19H17NO6S
MDL Number MFCD03792475
CAS 197245-19-5
Melting Point 167° to 169°C
Beilstein 8282210
Quantity 5g
Synonym N-(Benzo[b]thiophenesulfone-2-methoxycarbonyl)-L-phenylalanine; Bsmoc-Phe-OH
Optical Rotation −36° (c=1 in DMF)
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C