Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Bsmoc-L-methionine, 99%

$185.66 - $185.66

Chemical Identifiers

Chemical Name or Material N-Bsmoc-L-methionine
CAS 197245-29-7
Molecular Formula C15H17NO6S2
Formula Weight 371.43g/mol
MDL Number MFCD03792474
Beilstein 8278161
Synonym N-(Benzo[b]thiophenesulfone-2-methoxycarbonyl)-L-methionine; Bsmoc-Met-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1974006

 
alfa aesar™
L19740-06
5g Each for $185.66
Description & Specifications

Specifications

Melting Point ≈116°C
Quantity 5g
Optical Rotation +7.9° (c=0.5 in Chloroform)
Chemical Name or Material N-Bsmoc-L-methionine
CAS 197245-29-7
Assay 99%
Molecular Formula C15H17NO6S2
Formula Weight 371.43g/mol
MDL Number MFCD03792474
Beilstein 8278161
Synonym N-(Benzo[b]thiophenesulfone-2-methoxycarbonyl)-L-methionine; Bsmoc-Met-OH
Recommended Storage 2° to 8°C