Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Bsmoc-L-asparagine, 98%

$166.61 - $166.61

Chemical Identifiers

Chemical Name or Material N-Bsmoc-L-asparagine
CAS 197245-31-1
Molecular Formula C14H14N2O7S
Formula Weight 354.34g/mol
MDL Number MFCD03792471
Beilstein 8282117
Synonym N-(Benzo[b]thiophenesulfone-2-methoxycarbonyl)-L-asparagine; N-Bsmoc-Asn-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1973606

 
alfa aesar™
L19736-06
5g Each for $166.61
Description & Specifications

Specifications

Melting Point 134° to 138°C
Quantity 5g
Optical Rotation −22° (c=1 in DMF)
Chemical Name or Material N-Bsmoc-L-asparagine
CAS 197245-31-1
Assay 98%
Molecular Formula C14H14N2O7S
Formula Weight 354.34g/mol
MDL Number MFCD03792471
Beilstein 8282117
Synonym N-(Benzo[b]thiophenesulfone-2-methoxycarbonyl)-L-asparagine; N-Bsmoc-Asn-OH
Recommended Storage 2° to 8°C