Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Bsmoc-L-alanine, 97%

$141.00 - $487.15

Chemical Identifiers

Chemical Name or Material N-Bsmoc-L-alanine
CAS 197245-15-1
Molecular Formula C13H13NO6S
Formula Weight 311.32g/mol
MDL Number MFCD03792470
Beilstein 8272137
Synonym N-(Benzo[b]thiophenesulfone-2-methoxycarbonyl)-L-alanine; Bsmoc-Ala-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1973503

 
alfa aesar™
L19735-03
1g Each for $141.00

AAL1973506

 
alfa aesar™
L19735-06
5g Each for $487.15
Description & Specifications

Specifications

Melting Point 105° to 107°C
Quantity 1g
Optical Rotation −18° (c=1 in DMF)
Chemical Name or Material N-Bsmoc-L-alanine
CAS 197245-15-1
Assay 97%
Molecular Formula C13H13NO6S
Formula Weight 311.32g/mol
MDL Number MFCD03792470
Beilstein 8272137
Synonym N-(Benzo[b]thiophenesulfone-2-methoxycarbonyl)-L-alanine; Bsmoc-Ala-OH
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C