Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Bsmoc-L-2-aminobutyric acid, 99%

Chemical Identifiers

Chemical Name or Material N-Bsmoc-L-2-aminobutyric acid
Molecular Formula C14H15NO6S
Formula Weight 325.34
MDL Number MFCD03792469
Synonym N-(Benzo[b]thiophenesulfone-2-methoxycarbonyl)-L-2-aminobutyric acid; Bsmoc-Abu-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1973403

 
alfa aesar™
L1973403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1973406

 
alfa aesar™
L1973406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 325.34
Chemical Name or Material N-Bsmoc-L-2-aminobutyric acid
Assay 99%
Molecular Formula C14H15NO6S
Melting Point 121° to 122°C
MDL Number MFCD03792469
Synonym N-(Benzo[b]thiophenesulfone-2-methoxycarbonyl)-L-2-aminobutyric acid; Bsmoc-Abu-OH
Optical Rotation −18° (c=1 in DMF)
Quantity 1g
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C