Learn More
N-Bromosuccinimide, 99%
CAS: 128-08-5 | C4H4BrNO2 | 177.99 g/mol
Supplier: Thermo Scientific Chemicals 107450010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent, C-X bond formation reagentSpecifications
| N-Bromosuccinimide | |
| 98.5 | |
| 175.0°C to 178.0°C | |
| 180°C | |
| Authentic | |
| Glass bottle | |
| MFCD00005510 | |
| 21, 380 | |
| 15, 1448 | |
| Solubility in water: 14.8g/L (20°C). Other solubilities: 3.10g/100g glacial acetic acid(25°C),g/100g solvent (25°C):,tert. butanol:0.73- aceton:14.40,carbone tetrachloride:0.02- hexane:0.006,benzene:0.5 | |
| C1CC(=O)N(C1=O)Br | |
| 177.99 | |
| CHEBI:53174 | |
| 99% |
| 128-08-5 | |
| 100.0 | |
| White to Yellow | |
| 1% max. | |
| 98.5 to 101.5% (on dry substance) (Iodometry) | |
| C4H4BrNO2 | |
| 1 kg | |
| 01,78; 02,40; 03,34; 04,49; 05,65; 06,74; 07,37; 08,54; 09,70; 10,57; 11,79; 12,79; 13,41; 14,57; 15,50; 16,49 | |
| n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide | |
| PCLIMKBDDGJMGD-UHFFFAOYSA-N | |
| 1-bromopyrrolidine-2,5-dione | |
| 67184 | |
| 177.99 | |
| Crystalline Powder |
Chemical Identifiers
| 128-08-5 | |
| PCLIMKBDDGJMGD-UHFFFAOYSA-N | |
| 67184 | |
| 1-bromopyrrolidine-2,5-dione |
| MFCD00005510 | |
| n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide | |
| CHEBI:53174 | |
| C1CC(=O)N(C1=O)Br |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
May be corrosive to metals.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
In case of major fire and large quantities: Evacuate area.
Fight fire rem
GHS Signal Word: Danger
EINECSNumber : 204-877-2
TSCA : TSCA
RUO – Research Use Only