missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Bromosaccharin 98.0+%, TCI America™
Supplier: TCI America B215225G
Specifications
N-Bromosaccharin | |
White-Yellow | |
MFCD03844772 | |
n-bromosaccharin, 2-bromobenzo d isothiazol-3 2h-one 1,1-dioxide, 2-bromo-1,2-benzisothiazol-3 2h-one 1,1-dioxide, 2-bromo-1??,2-benzothiazole-1,1,3-trione | |
BrN1C(=O)C2=CC=CC=C2S1(=O)=O | |
262.08 | |
262.08 | |
Crystalline Powder |
35812-01-2 | |
C7H4BrNO3S | |
25 g | |
QRADPXNAURXMSB-UHFFFAOYSA-N | |
2-bromo-2,3-dihydro-1λâ¶,2-benzothiazole-1,1,3-trione | |
64791 | |
≥98.0% (T) |
Chemical Identifiers
35812-01-2 | |
262.08 | |
QRADPXNAURXMSB-UHFFFAOYSA-N | |
64791 | |
BrN1C(=O)C2=CC=CC=C2S1(=O)=O |
C7H4BrNO3S | |
MFCD03844772 | |
n-bromosaccharin, 2-bromobenzo d isothiazol-3 2h-one 1,1-dioxide, 2-bromo-1,2-benzisothiazol-3 2h-one 1,1-dioxide, 2-bromo-1??,2-benzothiazole-1,1,3-trione | |
2-bromo-2,3-dihydro-1λâ¶,2-benzothiazole-1,1,3-trione |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator