missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(Bromomethyl)phthalimide 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 403070250

 View more versions of this product

Catalog No. AC403070250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
240.06
White to off-white
100.0
Glass bottle
97%
5332-26-3
N-(Bromomethyl)phthalimide
Crystalline Powder
25g
149° to 154°C
96.0
Authentic
MFCD00005897
97%
C9H6BrNO2

Chemical Identifiers

5332-26-3
100.0
C9H6BrNO2
MFCD00005897
≥96.0%
96.0
97%
240.06
N-(Bromomethyl)phthalimide, 97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE

EINECSNumber : 226-239-2

TSCA : TSCA