missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(Bromomethyl)phthalimide 97%, ACROS Organics™

Chemical Identifiers

CAS 5332-26-3
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H6BrNO2
Formula Weight 240.06
MDL Number MFCD00005897
Chemical Name or Material N-(Bromomethyl)phthalimide, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC403070250 SDS Thermo Fisher Scientific
403070250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

5332-26-3
100.0
C9H6BrNO2
MFCD00005897
≥96.0%
96.0
97%
240.06
N-(Bromomethyl)phthalimide, 97%

Specifications

5332-26-3
100.0
Glass bottle
White to off-white
25g
C9H6BrNO2
MFCD00005897
≥96.0%
96.0
Authentic
149° to 154°C
Crystalline Powder
97%
240.06
N-(Bromomethyl)phthalimide, 97%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE

EINECSNumber : 226-239-2

TSCA : TSCA