Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-trans-4-hydroxy-D-proline, 99%

$251.64 - $637.36

Chemical Identifiers

Chemical Name or Material N-Boc-trans-4-hydroxy-D-proline
CAS 147266-92-0
Molecular Formula C10H17NO5
Formula Weight 231.25
MDL Number MFCD01861341
Synonym Boc-D-Hyp-OH; 1-tert-Butoxycarbonyl-(2R,4S)-4-hydroxypyrrolidine-2-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2715503

  SDS
alfa aesar™
H2715503
1g Each for $637.36

AAH27155MD

  SDS
alfa aesar™
H27155MD
250mg Each for $251.64
Description & Specifications

Specifications

Formula Weight 231.25
Synonym Boc-D-Hyp-OH; 1-tert-Butoxycarbonyl-(2R,4S)-4-hydroxypyrrolidine-2-carboxylic acid
Melting Point ≈123°C
Recommended Storage 2° to 8°C
Assay 99%
CAS 147266-92-0
Chemical Name or Material N-Boc-trans-4-hydroxy-D-proline
Molecular Formula C10H17NO5
MDL Number MFCD01861341
Quantity 1g
Safety and Handling

2° to 8°C