Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-trans-4-(Fmoc-amino)-L-proline, 97%

Chemical Identifiers

Chemical Name or Material N-Boc-trans-4-(Fmoc-amino)-L-proline
CAS 176486-63-8
Formula Weight 452.5
MDL Number MFCD00673783
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6687803

 
alfa aesar™
H6687803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66878MD

 
alfa aesar™
H66878MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 197° to 201°C
Formula Weight 452.5
MDL Number MFCD00673783
Assay 97%
Chemical Name or Material N-Boc-trans-4-(Fmoc-amino)-L-proline
CAS 176486-63-8
Physical Form Powder
Quantity 1g
Color White/off-white/palebrown